Veggie-Bites™ Guinea Pig Enrichment Treats, Certified
Catalog #: CA89067-444
Supplier: Bio-Serv
Specifications
- Description:Veggie-bites, certified
- Packaging:2 × 5 kg Bags/box
- Supplier No.:F5158
Specifications
About this item
An all-natural, crunchy vegetable treat that is a medley of dehydrated sliced carrots, puffed peas and whole kernel corn.
- Less labor intensive and more sanitary than perishable vegetables
- Feed as a training reward or foraging treat
- Nutritionally assayed
- Shelf life: 12 months, cool dry storage
Ingredients: Dehydrated carrots, puffed peas, dried whole kernel corn.
Caloric value: 3.44 kcal/gm
Nutritional ash content: 2.0%
Nutritional carbohydrate content: 67.0%
Nutritional fat content: 4.0%
Nutritional fibre content: 5.0%
Nutritional moisture content: <12%
Nutritional protein content: 10.0%
Certifications: Certified (Contaminant screened).
Ordering information: Product is no longer gamma irradiated. It is now Certified. If gamma irradiation is required, please contact customer service for pricing and lead time.
Customers Who Bought This Also Bought