Transgenic Dough Diet™ for Rodents, Sterile, Bio-Serv
Catalog #: CA89067-442
Supplier: Bio-Serv
Specifications
- Description:Transgenic Dough, Bacon Flavor
- Cat. no.:CA89067-442
- Packaging:1 kg/Bag
- Supplier No.:S3472
Specifications
About this item
Nutritionally complete, soft diet with higher protein and fat levels to comply with NRC and Transgenic-users' recommendations
- Designed for rodents with chewing, dental or mobility impairments
- Cookie dough consistency replaces labor intensive, nutrient-diluted mush diets
- Ideal for recovering rodents and weanlings or any time a soft diet is needed
- Rich bacon flavor stimulates appetite in dogs and rodents
- Place in feeder cup or on cage floor
- Sterilized by gamma irradiation
- Nutritionally assayed
- Shelf Life: 6 months refrigerated, 12 months frozen
Ingredients: Sucrose, Whole Milk Powder, Casein, Soybean Oil, Cellulose, Corn Syrup, Vitamin Mix, Mineral Mix, Glycerin, Bacon Flavor, DL-Methionine, Choline Bitartrate, Phenylalanine, tBHQ.
Caloric value: 3.83 kcal/g
Nutritional ash content: 3.3%
Nutritional carbohydrate content: 46.5%
Nutritional fat content: 12.4%
Nutritional fibre content: 4.8%
Nutritional moisture content: <12%
Nutritional protein content: 21.2%
Customers Who Bought This Also Bought