KleenGuard™ A70 Chemical Spray Protection Coveralls, Kimberly-Clark Professional
Catalog #: CA89030-826
Supplier: Kimberly-Clark
Specifications
- Anti-static:Yes
- Clothing Size:XL
- Garment application:Industrial,Chemical protection,Laboratory
- Color:Yellow
- Description:Elastic Wrists, Ankles, and Hood
- Seam type:Bound
- Cat. no.:CA89030-826
- Supplier No.:09814
Specifications
About this item
Manufactured from NFPA 99 compliant antistatic material, coveralls feature a seamless front for superior protection, ample chest room and more body length, and a liquid-resistant, extra-long zipper
- For use in hazardous waste remediation, environmental cleanup, petrochemical/oil refining, acid/caustic handling, biohazard cleanup, and pharmaceutical manufacturing applications
- Features bound seams for increased protection
- Durable, breathable, and comfortable
- Manufactured from NFPA 99 compliant antistatic material
- Wick-away lining provides enhanced comfort during extended use
The KleenGuard™ A70 line of apparel is designed for use in hazardous waste remediation, environmental cleanup, petrochemical/oil refining, acid/caustic handling, biohazard cleanup, and pharmaceutical manufacturing applications. Apparel features bound seams for increased protection. The soft and quiet fabric is durable, breathable, and comfortable.
Wick-away lining provides enhanced comfort during extended use, and a storm flap covers the zipper for additional protection. Available with a variety of hood and boot configurations. Level B/C suit.
Certifications: Passes NFPA 99 for antistatic materials.
Customers Who Bought This Also Bought