Specifications
- Clothing Size:L
- Garment application:Laboratory
- Color:White
- Description:Hooded Coveralls with Elastic Ankles
- Cat. no.:CA89012-814
Specifications
About this item
Coveralls provide high levels of protection and durability while remaining comfortable. A unique flash spinning process creates an excellent barrier against dry particulates, nonhazardous liquids, and microorganisms.
Coveralls provide high levels of protection and durability while remaining comfortable. A unique flash spinning process creates an excellent barrier against dry particulates, nonhazardous liquids, and microorganisms. Antistatic coveralls are low-linting and chemically and biologically inert.
- Protective, durable, and comfortable
- Excellent barrier against dry particulates, nonhazardous liquids, and microorganisms
- Chemically and biologically inert, low linting, and static free until laundered
- Serged seams, front zipper closure, elastic wrists, and attached hood
- Available with elastic ankles, or with attached boots that feature polyethylene anti-skid soles
Antistatic coveralls are low-linting and chemically and biologically inert.
Antistatic coveralls are low-linting and chemically and biologically inert.
All coveralls feature serged seams, front zipper closure, elastic wrists, and attached hood. Available with elastic ankles, or with attached boots that feature polyethylene anti-skid soles.