Specifications
- W×L:31.8 x 34 cm (121/2 x 133/8")
- Color:White
- Cat. no.:CA89004-556
- Packaging:Jumbo Roll
- Supplier No.:41025
Specifications
About this item
Wipers absorb oil and water faster than rags or laundered towels.They maintain strength even in solvents. Long-lasting, on-the-job performance extends usage, reducing cost and waste.
Durable and economical alternative to laundered shop towels and rags. Suitable for a wide range of laboratory and industrial tasks, including oil, solvent, and chemical cleanup. Wipers absorb oil and water faster than rags or laundered towels. They provide consistent size and strength without the inconvenience of holes, strings, or residues.
Constructed of HYDROKNIT™, a hydroentangled material manufactured without adhesives or chemical binders. Reusable wipers will not scratch skin or delicate surfaces, and will not fall apart when wet. They maintain strength even in solvents. Long-lasting, on-the-job performance extends usage, reducing cost and waste.
- Durable and economical alternative to laundered shop towels and rags
- Suitable for a wide range of laboratory and industrial tasks, including oil, solvent, and chemical cleanup
- Consistent size and strength without the inconvenience of holes, strings, or residues
- HYDROKNIT® hydroentangled material manufactured without adhesives or chemical binders will not scratch skin or delicate surfaces
- Reusable towels will not fall apart when wet