To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:540 ml (18 oz.)
- W×L:114×229 mm (4.5×9")
- Thickness:3 mil
- Cat. no.:CA89003-818
Specifications
About this item
These sterile sampling bags are ideal for collecting chlorinated water specimens.
- Sterile polyethylene construction
- Suited for water sampling
- Easy to use
- Wire closure provides an airtight seal
They are made of sterile virgin polyethylene tubing with no side seals.
Bags feature a wire closure for an airtight seal and wide-sealed bottoms for added leakproof protection. They are easy to use: simply tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.
UPA-approved for drinking water analysis.
Bags include a nontoxic, nutritive tablet containing 10 mg of active sodium thiosulfate.