Specifications
- Color:Green
- Description:Biopsy Cassettes
- Cat. no.:CA87002-400
- Supplier No.:M486-4SL
Specifications
About this item
Save time and money with convenient sleeved cassettes.
- Suitable for hoppers accepting plastic sleeves in labeling instruments
- Cassettes are resistant to chemical action of histological solvents
- Efficient flow-through slots maximize fluid exchange and ensure proper draingage
Specially designed cassettes are molded from a special high density polymer designed to keep specimens safely submerged in liquid.
Anterior printing area is a 45 degree angle, offering an unobstructed view of the writing surface and making the cassette perfectly suitable to be used with cassette labeling instruments. The one piece disposable plastic cover can be opened and closed as often as necessary and it always relocks without danger of specimen loss.
Each case contains 10 sleeves and 750 covers (75 sleeves per tube).
Available in both tissue and biopsy cassettes. The biopsy model is designed to hold small biopsy samples securely during the embedding process.