VWR® Centrifugal Filters
Catalog #: CA82031-344
Supplier: VWR International
Specifications
- Length:4.5 cm (125/32")
- Color:Gray
- Description:Polyethersulfone (PES) membrane
- Operating temperature range:0...40 °C (32...104 °F)
- Max. sample volume:500 µl
- Hold-up volume:5 µl
- Effective filtration area:0.28 cm2
- Max. centrifugal force:14000 g
- Max. receiver volume:500 µl
- Cat. no.:CA82031-344
- MWCO:3K
Specifications
About this item
Use to: Concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA; clean up labeling and PCR reactions; isolate DNA from agarose gel slices; buffer exchange; separate proteins, oligonucleotides, and RNA from acrylamide gels; clarify samples; or remove particulates.
- Polypropylene housing
- Membrane options available
Filter housings are constructed of polypropylene and are available with a polyethersulfone (PES) or nylon membrane.
Filters with nylon membrane are ideal for prefiltration clean-up of large agarose or acrylamide gel debris prior to further purification with an MWCO device.
Customers Who Bought This Also Bought