ContactUsLinkComponent
CHAPS Zwitterionice Detergents
Zwitterionic Detergent Solutions, G-Biosciences
Catalog #: CA82023-016
Supplier:  G-Biosciences
Zwitterionic Detergent Solutions, G-Biosciences
Catalog #: CA82023-016
Supplier:  G-Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    CHAPS
  • Size:
    5 g
  • Cat. no.:
    CA82023-016
  • Supplier No.:
    DG050
Specifications
About this item
Zwitterionic detergents protect the native state of proteins without altering the native charge of the protein molecules
G-Biosciences Zwitterionic Detergent Solutions include: CHAPS, CHAPSO, Sulfobetaine 3-10 (SB 3-10), Sulfobetaine 3-12 (SB 3-12), Sulfobetaine 3-14 (SB 3-14), ASB-14, ASB-16, and ASB-C8Ø.
Zwitterionic detergents are used for isoelectric focusing and 2D electrophoresis. Synthetic zwitterionic detergents are known as sulfobetaines. Sulfobetaines retain their zwitterionic characteristics over a wide range of pH.
Ordering information: Accessories not included.
Customers Who Bought This Also Bought