WYPALL® Waterless Cleaning Wipes, KIMBERLY-CLARK PROFESSIONAL®
Catalog #: CA82017-957
Supplier: Kimberly-Clark
Specifications
- Color:Green/White
- Description:Waterless Cleaning Wipes
- Cat. no.:CA82017-957
- Packaging:50 Wipes/Canister
- Supplier No.:58310
Specifications
About this item
Waterless wash-up is easy with Waterless Cleaning Wipes, particularly for on-the-go maintenance and cleaning tasks.
Waterless wash-up is easy with Waterless Cleaning Wipes, particularly for on-the-go maintenance and cleaning tasks.
These pre-moistened wipes serve as a heavy-duty waterless cleanser. Orange peel extract cleaning agent. Alcohol-free formulation, with soothing ingredients that aid in moisturizing skin.
Customers Who Bought This Also Bought