KLEENGUARD® A40 Liquid and Particle Protection Lab Coats, Kimberly-Clark Professional®
Catalog #: CA82007-624
Supplier: Kimberly-Clark
Specifications
- Clothing Size:3XL
- Garment application:Chemical protection
- Color:White
- Environmentally Preferable:Waste Reducing
- Cat. no.:CA82007-624
- Supplier No.:44446
Specifications
About this item
Manufactured from a breathable microporous film laminate, lab coats provide an excellent particle barrier and resist nonhazardous liquids
Meets ANSI/ISEA 101-1996. Passes ASTM F1670 testing for penetration of blood and body fluids. NFPA compliant antistatic material.
These low-linting garments are ideally suited for use in manufacturing, cleaning, utility, liquid handling, pressure washing, and maintenance applications. The tough exterior film protects while its soft nonwoven fabric interior provides comfort against the skin. KLEENGUARD® A40 laminate has a dry particle holdout of 100% between 0.3–0.5µm.
Customers Who Bought This Also Bought