KleenGuard™ A40 Liquid and Particle Protection Coveralls, Kimberly-Clark Professional
Catalog #: CA82007-584
Supplier: Kimberly-Clark
Specifications
- Anti-static:Yes
- Clothing Size:L
- Garment application:Industrial,Chemical protection,Laboratory
- Color:White
- Description:Elastic Wrists, Ankles, and Hood
- Environmentally Preferable:Waste Reducing
- Model:A40
- Cat. no.:CA82007-584
- Supplier No.:44323
Specifications
About this item
KleenGuard™ A40 Coveralls are designed to be used in applications where Tyvek® garments are typically worn, and provide an excellent particle barrier and resist nonhazardous liquids.
- Zippered front
- Breathable white microporous film keeps out debris, non-hazardous liquid chemicals and dry particulates
- Provides a better liquid barrier and a better particulate barrier than DuPont Tyvek®
- Pass NFPA 99 for antistatic materials
- Pass ASTM F1670 testing for penetration of blood and bodily fluids
- Meet the ANSI/ISEA 101-1996 sizing standards
KleenGuard™ A40 Liquid & Particle Protection Coveralls feature a zippered front. Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates, the KleenGuard™ A40 material provides employees with a better liquid barrier and a better particulate barrier than DuPont Tyvek®.
Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing, and many more.
Certifications: Pass NFPA 99 for antistatic materials; pass ASTM F1670 testing for penetration of blood and bodily fluids; and meet the ANSI/ISEA 101-1996 sizing standards.
Ordering information: Coveralls 89005-054 and -056 are individually wrapped.
Customers Who Bought This Also Bought