ContactUsLinkComponent
D-TUBE DIALYZER MIDI MWCO 6-8
D-Tube™ and D-Tube96™ Dialyzers, MilliporeSigma
Catalog #: CA80602-908
Supplier:  MilliporeSigma
D-Tube™ and D-Tube96™ Dialyzers, MilliporeSigma
Catalog #: CA80602-908
Supplier:  MilliporeSigma
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Volume:
  • Description:
    D-Tube™ Dialyzer Midi
  • Size:
    10 Tubes
  • Cat. no.:
    CA80602-908
  • MWCO:
    6 - 8 kDa
  • For Sample Range:
    50 - 800 µl
  • Supplier No.:
    71507-3
Specifications
About this item
D-Tube Dialyzers are easy to handle tubes with dual membranes providing a large surface area for fast, efficient dialysis and electroelution of proteins, RNA, DNA, and oligonucleotides from polyacrylamide or agarose gels.
  • Efficient dialysis - dual membrane surface
  • Convenient - use of single or multichannel pipet or robotic dispensing
  • High sample recovery, >97%
  • Protease-, RNase-, and DNase-free
  • Versatile - dialyze proteins, oligonucleotides, RNA, or DNA
  • Ideal for buffer exchange and removal of detergents and urea
The membrane is ultra clean, EDTA-treated, regenerated cellulose that is sulfur- and heavy metal-free. D-Tube96™ Dialyzers allow convenient, high throughput dialysis of 96 samples simultaneously.
Customers Who Bought This Also Bought