Order Entry
ContactUsLinkComponent
Benzonase Nuclease >90% 2.5Ku
Benzonase® Nuclease
Catalog #: CA80600-380
Supplier:  MilliporeSigma
CAS Number:  
Benzonase® Nuclease
Catalog #: CA80600-380
Supplier:  MilliporeSigma
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    2.5 ku
  • Enzyme name:
    Benzonase® nuclease
  • Purity:
    >90%
  • Concentration:
    25-29 U/µL
  • Cat. no.:
    CA80600-380
  • Supplier No.:
    70746-4

Specifications

About this item

Effective viscosity reduction and removal of nucleic acids from protein solutions.

  • The 99% purity grade is tested for endotoxins and contains 0.25 EU/1000 units
  • It is supplied as a 0.2 µm filtered solution in 50% glycerol
  • These are genetically engineered endonuclease from Serratia marcescens
  • Stores at –20 °C

Benzonase Nuclease is available in ultrapure and pure grades at a standard concentration of 25-29 U/µL and at a high concentration (HC) of 250 U/µL. Both preparations are free of detectable protease and have specific activity 1 × 10⁶ U/mg protein. It degrades all forms of DNA and RNA (single stranded, double stranded, linear and circular) while having no proteolytic activity. It is effective over a wide range of conditions and possesses an exceptionally high specific activity.

Customers Who Bought This Also Bought