ContactUsLinkComponent
APRON TVK 28X36BIB BOUND CS100
DuPont™ Tyvek® 400 Bib Aprons
Catalog #: CA80080-916
Supplier:  DuPont
APRON TVK 28X36BIB BOUND CS100
DuPont™ Tyvek® 400 Bib Aprons
Catalog #: CA80080-916
Supplier:  DuPont
Specifications
  • Garment application:
    Industrial,
    Paint and lacquer
  • Color:
    White
  • Cat. no.:
    CA80080-916
  • Supplier No.:
    TY273BWH0000
Specifications
About this item
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Neck loop
  • Waist ties
  • Bound seams
  • One size fits most
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include: lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
These bib aprons provide enhanced frontal protection. Bound seams and ties at waist allow for one size to fit most.
Certifications: Thickness meets ASTM D1777.
Customers Who Bought This Also Bought