DuPont™ Tyvek® IsoClean® Coveralls with Attached Hood
Catalog #: CA80077-176
Supplier: DuPont
Specifications
- Clothing Size:XL
- Garment application:Cleanroom,Laboratory
- Color:White
- Cat. no.:CA80077-176
- Chest Size:271/4"
- Inseam:291/2"
- Supplier No.:IC180SWHXL00
- Sleeve:361/2"
Specifications
About this item
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
- Serged seams
- Attached elastic hood
- Set sleeve design
- Elastic wrists and ankles
- Zipper closure
Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls have elastic openings for a tighter fit at wrists and ankles. Attached hood with elastic around face opening ensures a secure fit. Front zipper closure allows for easy donning and doffing.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Customers Who Bought This Also Bought