Specifications
- Clothing Size:4XL
- Garment application:Cleanroom,Laboratory
- Sterile:+
- Color:White
- Cat. no.:CA80077-048
- Chest Size:38"
- Inseam:35"
- Supplier No.:IC182BWH4XCS
- Sleeve:42"
Specifications
About this item
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
Clean-processed garments are processed to minimize particle shedding and individually packaged in an ISO Class 4 cleanroom. Sterile garments are gamma sterilized to a Sterility Assurance Level (SAL) of 10-6.
- Bound seams
- Bound neck
- Raglan sleeve design
- Covered elastic wrists and ankles
- Zipper closure
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration. Coveralls have elastic openings at wrists and ankles for a tighter fit. Bound neck ensures lower particle shedding. Front zipper closure allows for easy donning and doffing. Raglan sleeve design allows for greater range of motion and comfort.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.