5-Bromo-4-chloro-3-indolyl-a-D-glucopyranoside
Catalog #: CA80050-468
Supplier: MilliporeSigma
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
214,30 $ to 2 349,04 $
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Ethanol denatured (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
147,27 $ to 464,44 $
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
500,19 $ to 1 790,36 $