ContactUsLinkComponent
Hemoccult Ii Dspnsapk Pk-100Kt
Hemoccult® Guaiac Fecal Occult Blood Test Systems, Beckman Coulter®
Catalog #: CA74020-016
Supplier:  Beckman Coulter
CAS Number:  
Hemoccult Ii Dspnsapk Pk-100Kt
Hemoccult® Guaiac Fecal Occult Blood Test Systems, Beckman Coulter®
Catalog #: CA74020-016
Supplier:  Beckman Coulter
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Hemoccult II® Dispensapak™ System
  • Cat. no.:
    CA74020-016
  • Supplier No.:
    61100
Specifications
About this item
Qualitative procedure for detecting fecal occult blood uses a guaiac paper slide test.
  • Qualitative
  • Pre-qualified
  • Lot to lot consistency
The rigorous pre-qualification of raw materials and a unique quality control system allows the manufacturing of lot to lot consistency that is unmatched in the industry.
Certifications: CLIA waived.
Ordering information: Single Slides pack 10012-000 is supplied with 100 slides, 100 applicator sticks, and two 15mL bottles of developer. Single Slides 80078-660 is packaged as a case that includes 1000 slides, 1000 applicator sticks, and twenty 15mL bottles of developer. Dispensapak™ system is supplied with 100 patient kits (triple slide and applicator sticks only) and six 15mL bottles of developer. Dispensapak™ Plus system is supplied with 40 patient kits and two 15mL bottles of developer. Patient Pak system includes 34 Hemoccult II® triple slides (test cards), two 15mL bottles of developer, and 102 applicator sticks. Hemoccult® Tape includes two tape dispensers and two 15mL bottles of developer.
Customers Who Bought This Also Bought