Two-Position Snap Caps, Simport Scientific
Catalog #: CA73960-344
Supplier: Simport Scientific
Specifications
- Color:Natural
- Description:Two-Position Snap Caps
- Cat. no.:CA73960-344
- For Tubes:17 mm Polystyrene Tubes
- Supplier No.:T401-10DSPE
Specifications
About this item
Dual position caps are suitable for use in both aerobic and anaerobic procedures.
- Cap for aerobic and anaerobic procedures
- For use with 12 and 17 mm tubes
- Polystyrene and polypropylene options available
Caps offer two possibilities. The closed but unsealed position where samples are maintained aerobic for microbiological procedures; and the fully sealed position where the cap is pushed tight to seal the tube for anaerobic use or for storage, transfer and centrifuge applications. Not autoclavable.
Customers Who Bought This Also Bought