Nunc® 96-Well Optical Bottom Plates, Thermo Scientific
Catalog #: CA73521-030
Supplier: Thermo Fisher Scientific
Specifications
- Well Volume:0.4 mL
- Color:White
- Working Volume:50 - 200 µl
- Surface Treatment:Collagen I
- Supplier No.:152040
Specifications
About this item
Plates have a polystyrene/polymer base and feature flat-bottom wells for plate reader access
The Poly-D-Lysine and Collagen I coatings create a positive charge on the surface that ensures cell attachment, growth, and differentiation. Wells provide optimum clarity for viewing well contents. The plate footprint is compatible with standard equipment and automated systems.
Ordering information: Lid is included.
Customers Who Bought This Also Bought