Nunc® Lids for Microplates, Thermo Scientific
Catalog #: CA73520-148
Supplier: Thermo Fisher Scientific
Specifications
- Sterility:Non sterile
- Color:Clear
- Description:Universal Lid
- Height:8.34 mm
- Ext. dimensions:127×85 mm
- Supplier No.:250003
Specifications
About this item
Nunc® microplate lids offer sample protection from contamination and evaporation during assay processing, incubation, or storage.
All lids are made of polystyrene. The 96-well plate lids feature condensation rings, which inhibit cross-contamination, and an evaporation barrier, which reduces edge effects. The ultra-low profile LowBot™ lids feature an evaporation rim and side cut-outs for robotic handling and reading bar code labels. They are optimized for automation and handling on low-profile 384- and 1536-well plates. These lids are available with either cut-off corners for NUNC® version plates or in a universal format to fit any plate.
Customers Who Bought This Also Bought