Specifications
- Description:10 ml tube with swab, 5 ml Sanicult Sterile Diluent solution media fill
- Cat. no.:CA73250-020
- Supplier No.:B30U-S5
Specifications
About this item
Swab test kits are for hard-to-reach places, surfaces, viscous liquids, semi-solids, and powders.
Swab test kits are for hard-to-reach places, surfaces, viscous liquids, semi-solids, and powders.
Ideal for applications in food and beverage, engineering, and petrochemical industries.
The kit consists of a sterile swab and buffered solution. To use, remove swab from diluent vial, swab sample area, return swab to vial, and mix well. The sample is easily stored and readily transported. Choose from 5 ml or 8.8 ml diluent solution volumes.