ContactUsLinkComponent
Sanicult* Sterile Swab and Vial System
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Catalog #: CA73250-017
Supplier:  Starplex
Sanicult* Sterile Swab and Vial System
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Catalog #: CA73250-017
Supplier:  Starplex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    10 ml tube with swab, 8.8 ml Sanicult Sterile Diluent solution media fill
  • Cat. no.:
    CA73250-017
  • Supplier No.:
    B30U-S
Specifications
About this item
Swab test kits are for hard-to-reach places, surfaces, viscous liquids, semi-solids, and powders.
Swab test kits are for hard-to-reach places, surfaces, viscous liquids, semi-solids, and powders.
Ideal for applications in food and beverage, engineering, and petrochemical industries.
The kit consists of a sterile swab and buffered solution. To use, remove swab from diluent vial, swab sample area, return swab to vial, and mix well. The sample is easily stored and readily transported. Choose from 5 ml or 8.8 ml diluent solution volumes.
Customers Who Bought This Also Bought