FORCEPS SPLNTR STRAIT 4.5IN
Catalog #: CA70150-008
Supplier: Almedic Limited
Customers Who Bought This Also Bought
Diethyl ether, anhydrous ≥99.0% (by GC) stabilized, GR ACS, Supelco®
Supplier: MilliporeSigma
$1,880.98 to $6,888.30
Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents, Thermo Scientific
Supplier: Thermo Fisher Scientific
$180.89
Sodium thiosulfate standardised solution 0.01 N in aqueous solution
Supplier: Thermo Scientific Chemicals
$69.00
Sol-Vet® Blood Collection Needles (Veterinary Care), Sol M
Supplier: SOL-MILLENNIUM MEDICAL
$16.24 to $318.45
Medium, Middlebrook 7H10, deep fill, 15x100mm plate, For the cultivation of Mycobacterium species
Supplier: HARDY DIAGNOSTICS CA
$106.15