Specifications
- Description:Antifoam A
- Size:100 mL
- Cat. no.:CA700013-438
- Packaging:Poly Bottle
- Supplier No.:A1302-100ML
Specifications
About this item
Antifoam A is also known as polydimethylsiloxane and is part of a group of polymeric organosilicon compounds known as silicones. It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.