ContactUsLinkComponent
Omnitray Single-Well Plate with Lid
Nunc® OmniTray Single-Well Plates with Lids, Thermo Scientific
Catalog #: CA62409-600
Nunc® OmniTray Single-Well Plates with Lids, Thermo Scientific
Catalog #: CA62409-600
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Non-Treated Surface
  • Supplier No.:
    242811
Specifications
About this item
Designed for many applications, including high throughput screening
These polystyrene, single-well plates have the same footprint as a 96-well plate. They also hold a 11.4x7.3cm (41/2x27/8") nylon membrane used for dot blotting bacterial clones. Lid protects cells from contamination while providing a means for cell respiration during incubation and/or storage. Radiation sterilized.
Nunclon™Δ certified surface ensures cell attachment and growth. MaxiSorp™ surface is ideal for arraying molecules with mixed hydrophilic/hydrophobic domains, such as proteins, directly onto the plate. Plate featuring an enhanced rectangular area is ideal for automatic picking and viewing systems.
Plates are useful for screening DNA libraries in high volume laboratories using robotics. They are also suitable for manual plating and screening. Compatible with Beckman Coulter®, Biomek®, Hybaid, Orca, Tecan® Trak, and Zymark® robotic systems.
Ordering information: Plates packaged 10 sleeves per case.
Customers Who Bought This Also Bought