To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Sterility:Non sterile
- Color:Clear
- Description:Standard Lid with Long Side Cut-Off Corner, Condesation Ring, and Evaporation Barrier
- Height:7.9 mm
- Ext. dimensions:127×85 mm
- Supplier No.:263339
Specifications
About this item
Nunc® microplate lids offer sample protection from contamination and evaporation during assay processing, incubation, or storage.
All lids are made of polystyrene. The 96-well plate lids feature condensation rings, which inhibit cross-contamination, and an evaporation barrier, which reduces edge effects. The ultra-low profile LowBot™ lids feature an evaporation rim and side cut-outs for robotic handling and reading bar code labels. They are optimized for automation and handling on low-profile 384- and 1536-well plates. These lids are available with either cut-off corners for NUNC® version plates or in a universal format to fit any plate.