Specifications
- Well Volume:0.3 mL
- Cat. no.:CA62409-116
- Well Shape:U-Bottom
- Supplier No.:262162
Specifications
About this item
These sterile, polystyrene plates are ideal for bacteriology, serology, molecular biology, and other laboratory applications
Plates have a standard format, which makes them compatible with plate washers and readers. V-bottom and U-bottom plates allow pelleting or settling of cells for gentle concentration and sample retrieval. Flat-bottom wells can be easily examined microscopically.
The wells have raised rims to reduce risk of cross-contamination. Lids have rings to control condensation during incubation. The beveled corners on the plates and lids prevent accidental reversal of the lid onto the plate. Wells are alphanumerically-labeled for easy identification. They stack conveniently and securely, with or without lids. Clone selection and replication can be performed using these plates with a few simple movements using a transferable solid phase (TSP) replicator.
Used for dilutions and general storage.