Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
Catalog #: CA62409-068
Supplier: Thermo Fisher Scientific
Specifications
- Well Volume:0.4 mL
- Color:Clear
- Material:Polystyrene
- Cat. no.:CA62409-068
- Packaging:5 Plates/Sleeve
- Well Shape:Flat-Bottom
- Supplier No.:269620
Specifications
About this item
These nonsterile 96-well plates are ideal for assays, colorimetric serology, and bacteriology applications
All plates are made of high-quality polystyrene to ensure optimum optical clarity, with the exception of plate 62409-108, which is made of polypropylene. Plate 260844 is generic bar coded.
Ordering information: Plate 62407-166 is provided with a lid. For lids and sealing tapes, see the 62409-118 series.
Customers Who Bought This Also Bought