Nunc® MicroWell™ 96-Well Plates, Polypropylene, High Volume, Thermo Scientific
Catalog #: CA62408-948
Supplier: Thermo Fisher Scientific
Specifications
- Sterility:Non sterile
- Well Volume:500 µL
- Color:Natural
- Supplier No.:267245
Specifications
About this item
Polypropylene plates have a standard 96-well format and are designed to fit standard automation equipment and robotics.
Black plates reduce background fluorescence and minimize back-scattered light.
White plates offer maximum signal reflection and minimize autoluminescence and autofluorescence signals.
Natural plates are ideal for screening, liquid phase assays and storage. They are also useful for combinatorial chemistry applications.
Blue, red, yellow, and natural plates exhibit minimum binding for hydrophilic molecules. They are ideal for sample collection and storage, library storage, and combinatorial chemistry applications.
Ordering information: Plates with round-bottom wells are supplied 10 per sleeve, 12 sleeves per case. Plates with conical-bottom wells are supplied 20 per sleeve, 6 sleeves per case. For lids and sealing films, see 73520-478 series. Bar coding is available; ask your sales representative.
Customers Who Bought This Also Bought