Order Entry
ContactUsLinkComponent
Nunclon* delta MultiDishes, Sterile
Nunclon™ Delta MultiDishes, Sterile, Thermo Scientific
Catalog #: CA62407-068
Nunclon* delta MultiDishes, Sterile
Nunclon™ Delta MultiDishes, Sterile, Thermo Scientific
Catalog #: CA62407-068

Specifications

  • Well Area:
    1.9 cm2
  • No. of wells:
    4
  • Packaging:
    4/Sleeve
  • External Dimensions:
    66 x 66 mm
  • Working Well Volume:
    0.4 - 1.0 ml
  • Supplier No.:
    176740

Specifications

About this item

These polystyrene dishes with flat-bottom wells incorporate the Nunclon™Δ treated surface for optimal cell attachment and growth

Lids are keyed for a one-way fit. Rings in the lids reduce evaporation and condensation. Writing areas on both lids and dishes and individual alphanumeric markings allow easy identification of samples. Nontoxic and certified nonpyrogenic. Stackable.

Customers Who Bought This Also Bought