Corning® Primaria™ Culture Plates, Corning
Catalog #: CA62406-455
Supplier: Corning
Specifications
- Sterility:Sterile
- Description:6-Well
- Material:PS
- Cat. no.:CA62406-455
- Packaging:1/Tray
- Supplier No.:353846
Specifications
About this item
Unique nitrogen-containing tissue culture surface chemistry. Improves attachment, spreading, and growth for many primary cells or cell lines. Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process.
- Stable, permanent surface modification
- Optically clear
- No special storage required
- Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
- Packaged in red color-coded, peel-open, medical-style packages
- Sterilized by gamma irradiation
- Nonpyrogenic tested to less than 0.1 EU/ml
Customers Who Bought This Also Bought