Order Entry
ContactUsLinkComponent
HIVEG PEPTONE 500G
HiVeg™ Peptones and Media, HiMedia Laboratories
Catalog #: CA61001-508
Supplier:  HiMedia
CAS Number:  
HiVeg™ Peptones and Media, HiMedia Laboratories
Catalog #: CA61001-508
Supplier:  HiMedia
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    HiVeg™ Peptone
    Growth Promotion Equivalent to Peptone
  • Size:
    500 g
  • Cat. no.:
    CA61001-508
  • Application:
    Culture Media, Enzyme, Vaccine, and Antibiotic Production
  • Supplier No.:
    RM001V-500G

Specifications

About this item

The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based peptones and media designed to work in conjunction with or to replace standard animal-based protein sources.

  • 100% Vegetable Protein Source
  • Eliminate the Risk of BSE/TSE
  • Equivalent Performance with Animal-Based Peptones

The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based peptones and media designed to work in conjunction with or to replace standard animal-based protein sources. The products effectively eliminate BSE/TSE contamination concerns while providing results on par with animal-based products. The wide selection of peptones and media can meet the requirements of nearly any microbiology application.

Customers Who Bought This Also Bought