Order Entry
ContactUsLinkComponent
BUSHNELL HAAS AGAR 500GM
Bushnell Haas Agar, HiMedia Laboratories
Catalog #: CA61000-080
Supplier:  HiMedia
CAS Number:  
Bushnell Haas Agar, HiMedia Laboratories
Catalog #: CA61000-080
Supplier:  HiMedia
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • pH:
    7.0 ±0.2
  • Description:
    Bushnell Haas Agar
  • Size:
    500 g
  • Storage Temperature:
    ≤ 30 °C
  • Supplier No.:
    M349-500G

Specifications

About this item

Bushnell Haas Agar are recommended for the microbial examination of fuels and for studying microbial hydrocarbon deterioration.

  • White to cream homogeneous free flowing powder
  • Firm gelling, comparable with 2.0% agar gel
  • pH: 6.80 to 7.20
  • Store below 30 °C

Bushnell Haas Agar contains magnesium sulfate (0.200 g/l); calcium chloride (0.020 g/l); monopotassium phosphate (1.000 g/l); dipotassium phosphate (1.000 g/l); ammonium nitrate (1.000 g/l); ferric chloride (0.050 g/l) and agar (20.000 g/l).

Customers Who Bought This Also Bought