To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Volume:1.5 ml
- Style:Conical Bottom
- Supplier No.:T341-5T
Specifications
About this item
Micrewtube™ Microcentrifuge tubes are ideal for freezer storage, boiling application and centrifugation, and will fit most standard microcentrifuge rotors.
- Made of polypropylene
- Can be used at temperatures from –196 to 121 °C
Low adhesion Micrewtubes™ Tubes are also available. The specially formulated polypropylene used to manufacture these tubes provides a low adhesion surface to obtain maximum sample yield.
Non-skirted tubes withstand high speed centrifugation up to 20000 ×g. Skirted tubes can withstand up to 17000 ×g. Available plain, graduated, or brown for light sensitive material. Available with or without molded ridges.