Order Entry
ContactUsLinkComponent
TUBE,MICREW2.0ML LP+O-RG CS500
Micrewtube® Microcentrifuge Tube with Screw Cap, Simport Scientific
Catalog #: CA60872-779
Supplier:  Simport Scientific
TUBE,MICREW2.0ML LP+O-RG CS500
Micrewtube® Microcentrifuge Tube with Screw Cap, Simport Scientific
Catalog #: CA60872-779
Supplier:  Simport Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Sterility:
    Sterile
  • Volume:
    2.0 ml
  • Cat. no.:
    CA60872-779
  • Style:
    Self Standing
  • Graduations:
    Graduated
  • Supplier No.:
    T332-6SPR

Specifications

About this item

MICREWTUBE® is ideal for freezer storage, boiling application and centrifugation.

The cap is molded with a deep internal lip that fits snugly against the interior wall of the tube thus preventing the contents from coming in contact with the seal or threads. In the silicone O-ring version, tubes and caps are made of polypropylene. Will withstand temperatures from –196 to +121 °C and are autoclavable. In the lip seal version, tubes are made of polypropylene and caps are made of high density polyethylene. Will withstand temperatures from –196 to +110 °C.

All tubes and closures are manufactured in a clean environment. Tubes and screw caps are made of heavy wall construction and are built to last. Non skirted tubes withstand high speed centrifugation up to 20000×g, skirted tubes up to 17000×g.

and will fit most standard microcentrifuge rotors. Six styles of caps to choose from and three sizes of conical bottom or self-standing tubes (0.5, 1.5 and 2 ml) are available in sterile and non sterile format. All sterile tubes are gamma irradiated and packaged in tamperproof, resealable bags to protect remaining tubes from contamination.

Certifications: Certified RNase, DNase, DNA and Pyrogen free.
Caution: Do not use Micrewtubes for storage in the liquid phase of liquid nitrogen. Such use may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build-up resulting in possible explosion or biohazard release. Use appropriate safety procedures when handling and disposing of vials.
Customers Who Bought This Also Bought