To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Length:37 mm
- Width:24 mm
- Height:10 mm
- Cat. no.:CA60872-023
- Supplier No.:M475-10
Specifications
About this item
These base molds offer the ease and convenience of a disposable mold, yet they are strong enough to be reused.
The specially formulated plastic material offers excellent thermal exchange. The molds have a smooth interior finish and rounded corners to facilitate specimen removal. They can be used with any standard cassettes and embedding rings.
The M475-10 base mold is designed especially for the M512 MACROSETTE®.