Order Entry
ContactUsLinkComponent
BD Falcon* Replacement Snap Cap for Test Tube
Falcon® Snap Caps for Test Tubes, Corning
Catalog #: CA60819-251
Supplier:  Corning
BD Falcon* Replacement Snap Cap for Test Tube
Falcon® Snap Caps for Test Tubes, Corning
Catalog #: CA60819-251
Supplier:  Corning

Specifications

  • Sterility:
    Sterile
  • Description:
    Falcon™ Snap Caps
  • Size:
    12 mm
  • Packaging:
    500/Pack
  • Fits:
    12×75 mm Test Tube
  • Supplier No.:
    352032

Specifications

About this item

Falcon® snap caps are designed for use with 12×75 mm test tubes

These polyethylene caps feature a patented, two-position fit, loose for aerobic culturing, tight for sealing. Sterile.

Ordering information: Snap caps are packed 500 per pack, 2000 per case.
Customers Who Bought This Also Bought