Order Entry
ContactUsLinkComponent
Safety Stand
Safety Stand for Overhead Stirrers, Caframo
Catalog #: CA58960-152
Supplier:  Caframo
Safety Stand
Safety Stand for Overhead Stirrers, Caframo
Catalog #: CA58960-152
Supplier:  Caframo

Specifications

  • Ø:
    25.4 mm
  • Length:
    710 mm
  • Description:
    Safety Stand for Overhead Stirrers
  • Cat. no.:
    CA58960-152
  • Supplier No.:
    A110

Specifications

About this item

The Caframo Safety Stand offers stable support for all Caframo stirrers.

The base is a cast zinc-aluminum alloy coated with chemical resistant epoxy paint and comes with two level adjustment knobs.

Ordering information: Includes the stand base and 28 x 1" dia. 304 stainless steel hollow rod. Additional rod lengths also available.
Customers Who Bought This Also Bought