Falcon® Disposable Aspirating Pipettes, Polystyrene, Sterile, Corning
Catalog #: CA53106-450
Supplier: Corning
Specifications
- Capacity:2 ml
- Cat. no.:CA53106-450
- Supplier No.:357558
Specifications
About this item
PS pipettes, non plugged, non graduated, disposable, sterile.
- Safer alternative to glass Pasteur pipettes
- Non pyrogenic
- Individually packed to prevent contamination
Packaging: Individually packed in a thermo-formed paper/plastic wrap.
Customers Who Bought This Also Bought