Order Entry
ContactUsLinkComponent
BD Falcon* CultureSlides*
Falcon® CultureSlides, Corning
Catalog #: CA53106-306
Supplier:  Corning
BD Falcon* CultureSlides*
Falcon® CultureSlides, Corning
Catalog #: CA53106-306
Supplier:  Corning

Specifications

  • Well Area:
    0.7 cm2
  • No. of wells:
    8
  • Working Volume:
    0.7 - 0.8 ml
  • Supplier No.:
    354118

Specifications

About this item

Soda-lime glass slide, PS vessel, lid and tool for loosening chamber.

  • Chamber easily and safely removed using the disposable safety removal tool
  • Pressure-sensitive, biocompatible, acrylic-adhesive gasket remains with the vessel after removal, not on the slide, facilitating further processing or placement of coverslips
  • Blue hydrophobic border defines cell culture areas
  • Wells numbered for easy identification
  • Trays designed for incubator use

Falcon® CultureSlides allow cells to be cultured and then analyzed on a glass microscope slide. Cells are grown in a plastic chamber attached to a specially prepared microscope slide. Cells can be fixed and stained in situ without disruption of the cell monolayer.

W×L: 25×75 mm with 1.2 mm bevelled edge

Ordering information: Supplied with removal tool and key.
Customers Who Bought This Also Bought