ContactUsLinkComponent
MINNCARE* Cold Sterilant
Minncare® Cold Sterilant, Mar Cor® Purification
Catalog #: CA50930-066
Minncare® Cold Sterilant, Mar Cor® Purification
Catalog #: CA50930-066
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    3.8 L (1 gal.)
  • Cat. no.:
    CA50930-066
  • Supplier No.:
    78399646
Specifications
About this item
Cold sterilant is a peracetic acid solution for use in a variety of medical, pharmaceutical, and industrial applications
Cold sterilant is a peracetic acid solution for use in a variety of medical, pharmaceutical, and industrial applications
It also provides protection for reverse osmosis membranes and associated distribution systems against short- and long-term bacterial problems. Solution provides 6 log reduction of bacteria population at a 100X dilution (1%) within a 36-minute soak time. Does not produce toxic aldehyde vapors. Biodegradable and easily disposable. Compatible with thin film composite membranes.
Customers Who Bought This Also Bought