Specifications
- Length:53 mm
- Volume:2 - 200 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Packed:960 Tips (10 trays × 96 Tips)
- Sterile:-
- Color Code:Yellow
- Supplier No.:022491539
Specifications
About this item
Eppendorf epT.I.P.S.® Reloads offer high-precision pipette tips in 'Eppendorf Quality' and 'PCR clean' purity grades. Eco-friendly, cost-effective, and designed for seamless compatibility with Eppendorf Box 2.0, they ensure accuracy and sustainability in labs.
- Reduced waste when compared with disposable racks
- Refill trays can be autoclaved up to 121 °C, in packaging and without foil, for subsequent use
- Depending on tip size, the refill system is packaged as either dual-sided or in stack form
- Available in purity grades of Eppendorf Quality and PCR clean
- Color-coded trays for simple volume and pipette identification
- ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf "Totally Integrated Pipetting System") pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.
How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.
Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.