Fetal bovine serum (FBS), premium grade, Corning®
Catalog #: CA45001-106
Supplier: Corning
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:FBS, Premium, US Sourced
- Hemoglobin:≤25 mg/dl
- Endotoxin:≤5.0 EU/ml
- Storage temperature:-25° to -15°C
- Grade:Premium
- Cat. no.:CA45001-106
- Supplier No.:35-015-CV
Specifications
About this item
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
- Sera is aseptically collected and processed in a cGMP facility
- Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
- Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
- Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
- Premium FBS is sourced from U.S. herds
- Heat-inactivated Fetal Bovine Serum (FBS) is heated at 56°C for 30 minutes
Customers Who Bought This Also Bought