Order Entry
ContactUsLinkComponent
L-ALANYL-GLUTAMINE
Corning® glutagro™ Supplement, Corning
Catalog #: CA45001-086
Supplier:  Corning
CAS Number:  
L-ALANYL-GLUTAMINE
Corning® glutagro™ Supplement, Corning
Catalog #: CA45001-086
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    glutaGRO™, Liquid 200 mM Solution (with 8.5 g/L NaCl)
  • Storage temperature:
    Ambient/Refrigerate
  • Shipping temperature:
    Ambient
  • Volume:
    100 ml
  • Supplier No.:
    25-015-CI

Specifications

About this item

Cells grown in traditional media supplemented with L-glutamine are susceptible to toxicity

With Corning® glutagro™, cells receive this needed supplement in the healthiest environment possible. This modified form of L-glutamine does not degrade into ammonia in storage and incubation, minimizing toxicity and improving culture quality.

Customers Who Bought This Also Bought