Order Entry
ContactUsLinkComponent
MEM NEAA 100X SOLN. 6X100ML
Corning® MEM (Minimum Essential Medium) Nonessential Amino Acids, Corning
Catalog #: CA45000-700
Supplier:  Corning
CAS Number:  
MEM NEAA 100X SOLN. 6X100ML
Corning® MEM (Minimum Essential Medium) Nonessential Amino Acids, Corning
Catalog #: CA45000-700
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    MEM Nonessential Amino Acids
  • Storage temperature:
    Frozen
  • Shipping temperature:
    Frozen
  • Volume:
    100 ml
  • Supplier No.:
    25-025-CI

Specifications

About this item

The addition of MEM nonessential amino acids to medium can improve growth and viability of propagating cells in culture while reducing the biosynthetic burden on these cells

This concentrated supplement allows for the efficient addition to media with no risk of dilution of media components.

Customers Who Bought This Also Bought