Order Entry
ContactUsLinkComponent
MCCOY'S 5A W/GLN 6X500ML
Corning® McCoy's 5A (Iwakata & Grace Modification), Corning
Catalog #: CA45000-374
Supplier:  Corning
CAS Number:  
MCCOY'S 5A W/GLN 6X500ML
Corning® McCoy's 5A (Iwakata & Grace Modification), Corning
Catalog #: CA45000-374
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    McCoy's 5A (Iwakata and Grace Modification) with L-glutamine
  • Storage temperature:
    Refrigerate
  • Cat. no.:
    CA45000-374
  • Supplier No.:
    10-050-CV

Specifications

About this item

McCoy's 5A medium was originally formulated to meet the culture requirements of Novikoff Hepatoma cells, and has been found to support the indefinite proliferation of Walker256 carcinoma cells along with a wide range of human and rat normal and transformed cells.  Originally modified by Iwakata and Grace, this version of McCoy's 5A includes increased levels of inositol and glucose

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
Customers Who Bought This Also Bought