Order Entry
ContactUsLinkComponent
ISCOVE'S W/HPS+GLN 6X500ML
Corning® DMEM, Iscove's Modification of DMEM, Corning
Catalog #: CA45000-366
Supplier:  Corning
CAS Number:  
ISCOVE'S W/HPS+GLN 6X500ML
Corning® DMEM, Iscove's Modification of DMEM, Corning
Catalog #: CA45000-366
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Iscove's Modification of DMEM with L-glutamine and 25 mM HEPES without ß-thioglycerol and ß-mercaptoethanol
  • Storage temperature:
    Refrigerate
  • Cat. no.:
    CA45000-366
  • Supplier No.:
    10-016-CV

Specifications

About this item

Iscove's Modification of DMEM (IMDM) is highly enriched synthetic medium optimized for rapidly proliferating cultures of high cell density

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

A modified form of Dulbecco's Modified Eagle Medium (DMEM), Iscove's contains additional amino acids, vitamins, selenium and HEPES buffer, and is formulated with potassium nitrate in place of ferric nitrate. The addition of albumin and transferrin may be required for certain cell types.

Customers Who Bought This Also Bought