Order Entry
ContactUsLinkComponent
IMEM RICHTER'S W/GLN 6X500ML
Corning® MEM (Minimum Essential Medium), Improved MEM (Richter's Mod.), Corning
Catalog #: CA45000-362
Supplier:  Corning
CAS Number:  
IMEM RICHTER'S W/GLN 6X500ML
Corning® MEM (Minimum Essential Medium), Improved MEM (Richter's Mod.), Corning
Catalog #: CA45000-362
Supplier:  Corning
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Improved MEM (Richter's Mod.) with L-glutamine
  • Storage temperature:
    Refrigerate
  • Cat. no.:
    CA45000-362
  • Supplier No.:
    10-024-CV

Specifications

About this item

A modified forumulation of Minimal Essential Medium, Improved Minimum Essential Medium (IMEM) contains a higher concentration of many components used in MEM along with additional amino acids, vitamins, and metabolites.

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment"
Customers Who Bought This Also Bought